Learn More
Medchemexpress LLC GSK1521498 free base 50mg | 1007573-18-3 | 50 MG
SDP
Catalog No. 5000226994
Encompass_Preferred
GSK1521498 free base is a potent, selective μ-opioid receptor (MOR) antagonist supplied for research use. It is used in neuroscience and pharmacology studies of opioid receptor-mediated behaviors, including compulsive consumption models. The material is provided as a solid with batch-specific analytical data.
  • Potent, selective μ-opioid receptor antagonist.
  • High analytical purity suitable for preclinical research.
  • Provided as solid powder for convenient handling.
  • Batch-specific certificate of analysis available.
  • Supplied in small pack sizes for dosing and assay development.
Catalog No. 50-002-26994 Supplier Medchemexpress LLC Supplier No. HY11506650MG
GSK1521498 free base is a potent, selective μ-opioid receptor (MOR) antagonist supplied for research use. It is used in neuroscience and pharmacology studies of opioid receptor-mediated behaviors, including compulsive consumption models. The material is provided as a solid with batch-specific analytical data.
  • Potent, selective μ-opioid receptor antagonist.
  • High analytical purity suitable for preclinical research.
  • Provided as solid powder for convenient handling.
  • Batch-specific certificate of analysis available.
  • Supplied in small pack sizes for dosing and assay development.