Learn More
Medchemexpress LLC HY-14348 5mg Medchemexpress, GSK163090 CAS:844903-58-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY143485MG
Encompass_Preferred
Medchemexpress, HY-14348 5mg GSK163090 CAS:844903-58-8 GSK163090 is a potent, selective and orally active 5-HT1A/1B/1D receptor antagonist with pKi values of 9.4/8.5/9.7, respectively. GSK163090 inhibits the functional activity of serotonin reuptake transporter (SerT) with a pKi value of 6.1. GSK163090 has antidepressant and anxiolytic activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1591