Learn More
Cayman Chemical GSK1702934A 10mg
SDP
Catalog No. 502947641
Encompass_Preferred
A TRPC3 and TRPC6 activator; induces TRPC3- and TRPC6-dependent currents in whole-cell patch-clamp assays using HEK293 cells expressing human channels (EC50s = 0.08 and 0.44 µM, respectively); enhances contractility and induces arrhythmias in hearts isolated from TRPC3-overexpressing mice
Catalog No. 50-294-7641 Supplier Cayman Chemical Supplier No. 3457610 MG
A TRPC3 and TRPC6 activator; induces TRPC3- and TRPC6-dependent currents in whole-cell patch-clamp assays using HEK293 cells expressing human channels (EC50s = 0.08 and 0.44 µM, respectively); enhances contractility and induces arrhythmias in hearts isolated from TRPC3-overexpressing mice