Learn More
Medchemexpress LLC HY-18990 2mg Medchemexpress, GSK180736A CAS:817194-38-0 Purity:>98%
SDP
Catalog No. 502030841
Encompass_Preferred
Medchemexpress, HY-18990 2mg GSK180736A CAS:817194-38-0 GSK180736A is potent Rho-associated coiled-coil kinase 1 (ROCK1) inhibitor with an IC50 of 100 nM. GSK180736A is also a selective and ATP-competitive G protein-coupled receptor kinase 2 (GRK2) inhibitor with an IC50 of 0.77 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0841 Supplier Medchemexpress LLC Supplier No. HY189902MG
Medchemexpress, HY-18990 2mg GSK180736A CAS:817194-38-0 GSK180736A is potent Rho-associated coiled-coil kinase 1 (ROCK1) inhibitor with an IC50 of 100 nM. GSK180736A is also a selective and ATP-competitive G protein-coupled receptor kinase 2 (GRK2) inhibitor with an IC50 of 0.77 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.