Learn More
Selleck Chemical LLC GSK180736A
SDP
Catalog No. 5000001413
Encompass_Preferred
GSK180736A (GSK180736) developed as a Rho-associated coiled-coil-containing protein kinase inhibitor binds to GRK2(G protein-coupled receptor kinase 2) with logIC50 pf -6 6 (logIC50 -4 0 for GRK5 and -3 for GRK1) 400-fold selective for GRK2 over both GRK1 and GRK5
Catalog No. 50-000-01413 Supplier Selleck Chemical LLC Supplier No. S848910MG
GSK180736A (GSK180736) developed as a Rho-associated coiled-coil-containing protein kinase inhibitor binds to GRK2(G protein-coupled receptor kinase 2) with logIC50 pf -6 6 (logIC50 -4 0 for GRK5 and -3 for GRK1) 400-fold selective for GRK2 over both GRK1 and GRK5