Learn More
Apexbio Technology LLC GSK1838705A 1116235-97-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062384
Encompass_Preferred
GSK1838705A (CAS 1116235-97-2) is a small molecule inhibitor targeting the kinase activity of insulin-like growth factor-1 receptor (IGF-1R) insulin receptor and anaplastic lymphoma kinase (ALK) It exhibits potent inhibition with IC50 values of 2 0 nM for IGF-1R 1 6 nM for insulin receptor and 0 5 nM for ALK In preclinical research GSK1838705A suppresses proliferation of hematologic malignancy-derived tumor cells including multiple myeloma and Ewing sarcoma lines and inhibits tumor xenograft growth in vivo Additionally despite targeting the insulin receptor it minimally affects glucose homeostasis at effective doses Due to its multi-target kinase inhibitory profile GSK1838705A serves as a promising tool for cancer biology research
Catalog No. 50-000-62384 Supplier Apexbio Technology LLC Supplier No. A84405.1
GSK1838705A (CAS 1116235-97-2) is a small molecule inhibitor targeting the kinase activity of insulin-like growth factor-1 receptor (IGF-1R) insulin receptor and anaplastic lymphoma kinase (ALK) It exhibits potent inhibition with IC50 values of 2 0 nM for IGF-1R 1 6 nM for insulin receptor and 0 5 nM for ALK In preclinical research GSK1838705A suppresses proliferation of hematologic malignancy-derived tumor cells including multiple myeloma and Ewing sarcoma lines and inhibits tumor xenograft growth in vivo Additionally despite targeting the insulin receptor it minimally affects glucose homeostasis at effective doses Due to its multi-target kinase inhibitory profile GSK1838705A serves as a promising tool for cancer biology research