Learn More
Medchemexpress LLC HY-14111 1mg Medchemexpress, GSK189254A CAS:720690-73-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY141111MG
Encompass_Preferred
Medchemexpress, HY-14111 1mg GSK189254A CAS:720690-73-3 GSK189254A (GSK189254) is a novel, potent and selective histamine H3 receptor antagonist with pKi values of 9.59-9.90 and 8.51-9.17 for human and rat H3, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1558