Learn More
Selleck Chemical LLC GSK1904529A 50mg 1089283-49-7 GSK 4529
SDP
Supplier:  Selleck Chemical LLC S109350MG
Encompass_Preferred
GSK1904529A (GSK 4529) is a selective inhibitor of IGF-1R and IR with IC50 of 27 nM and 25 nM in cell-free assays, >100-fold more selective for IGF-1R/InsR than Akt1/2, Aurora A/B,B-Raf, CDK2, EGFR etc. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-798-5