Learn More
TARGETMOL CHEMICALS INC GSK2008607 5MG
SDP
Catalog No. 502580612
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. GSK2008607 is a potent B-RafV600E inhibitor with anticancer activity and can be used to study breast colorectal melanoma thyroid and ovarian cancers. purity: 98%
Catalog No. 50-258-0612 Supplier TARGETMOL CHEMICALS INC Supplier No. T274545MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. GSK2008607 is a potent B-RafV600E inhibitor with anticancer activity and can be used to study breast colorectal melanoma thyroid and ovarian cancers. purity: 98%