Learn More
Medchemexpress LLC HY-19511 5mg Medchemexpress, GSK2018682 CAS:1034688-30-6 Purity:>98%
SDP
Catalog No. 502082137
Encompass_Preferred
Medchemexpress, HY-19511 5mg GSK2018682 CAS:1034688-30-6 GSK2018682 is an agonist for S1P1 and S1P5 receptor with pEC50s of 7.7 and 7.2, respectively, and has no agonist activity towards human S1P2, S1P3, or S1P4. GSK2018682 is used in the research of multiple sclerosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2137 Supplier Medchemexpress LLC Supplier No. HY195115MG
Medchemexpress, HY-19511 5mg GSK2018682 CAS:1034688-30-6 GSK2018682 is an agonist for S1P1 and S1P5 receptor with pEC50s of 7.7 and 7.2, respectively, and has no agonist activity towards human S1P2, S1P3, or S1P4. GSK2018682 is used in the research of multiple sclerosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.