Learn More
Medchemexpress LLC GSK215 | 2743427-26-9 | 99.3% | C50H59F3N10O6S | 10MG
SDP
Catalog No. 5000260979
Encompass_Preferred
GSK215 is a research-use PROTAC that selectively degrades focal adhesion kinase (FAK) by recruiting the VHL E3 ligase. It exhibits high potency (pDC50 ≈ 8.4) and causes rapid, prolonged reduction of FAK levels in cellular assays, making it useful for probing FAK-dependent signaling and pharmacology. Not for human or clinical use.
  • Selective FAK degradation via VHL E3 ligase.
  • High potency (pDC50 ≈ 8.4).
  • Rapid and sustained target knockdown in cells.
  • Supplied as a high-purity research compound.
  • Available in multiple milligram pack sizes for screening.
Catalog No. 50-002-60979 Supplier Medchemexpress LLC Supplier No. HY13229610MG
GSK215 is a research-use PROTAC that selectively degrades focal adhesion kinase (FAK) by recruiting the VHL E3 ligase. It exhibits high potency (pDC50 ≈ 8.4) and causes rapid, prolonged reduction of FAK levels in cellular assays, making it useful for probing FAK-dependent signaling and pharmacology. Not for human or clinical use.
  • Selective FAK degradation via VHL E3 ligase.
  • High potency (pDC50 ≈ 8.4).
  • Rapid and sustained target knockdown in cells.
  • Supplied as a high-purity research compound.
  • Available in multiple milligram pack sizes for screening.