Learn More
Medchemexpress LLC HY-132296 5mg , GSK215 CAS:2743427-26-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1322965MG
Encompass_Preferred
HY-132296 5mg GSK215 CAS: 2743427-26-9 GSK215 is a potent and selective PROTAC focal adhesion kinase (FAK) degrader with a pDC50 of 8.4. GSK215 is designed by a binder for the VHL E3 ligase and the FAK inhibitor VS-4718. GSK215 induces rapid and prolonged FAK degradation, giving a long-lasting effect on FAK levels and a marked pharmacokinetic/pharmacodynamics (PK/PD) disconnect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1752