Learn More
Cayman Chemical GSK2200150A, Purity: ≥98%, CAS: 1443138-53-1, 10 mg
SDP
Catalog No. 502913599
Encompass_Preferred
An antimycobacterial agent; active against M. tuberculosis and M. bovis (MICs = 0.38 and 1.6 µM, respectively)
Catalog No. 50-291-3599 Supplier Cayman Chemical Supplier No. 3662010 MG
An antimycobacterial agent; active against M. tuberculosis and M. bovis (MICs = 0.38 and 1.6 µM, respectively)