Learn More
Medchemexpress LLC HY-100183 50mg Medchemexpress, GSK2239633A CAS:1240516-71-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10018350MG
Encompass_Preferred
Medchemexpress, HY-100183 50mg GSK2239633A CAS:1240516-71-5 GSK2239633A is a CC-chemokine receptor 4 ( CCR4 ) antagonist, which inhibits the binding of [ 125 I]-TARC to human CCR4 with a pIC 50 of 7.96 ± 0.11. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1738