Learn More
Medchemexpress LLC GSK2245035 | 1207629-49-9 | 99.8% | 390.52 g/mol | C20H34N6O2 | 50 MG
SDP
Catalog No. 5000227680
Encompass_Preferred
GSK2245035 is a selective Toll-Like receptor 7 (TLR7) agonist for research use, preferentially stimulating type I interferon production and evaluated for intranasal administration in modulation of allergic responses. It is supplied as a high-purity, off-white to light-yellow solid suitable for laboratory studies.
  • Selective TLR7 agonist with preferential type I interferon stimulation.
  • High purity appropriate for in vitro and in vivo research.
  • Off-white to light-yellow solid form with defined storage stability.
  • Available as powder and as ready-to-use DMSO solutions for flexible dosing.
  • Commonly used in studies of innate immunity, antiviral responses, and allergic inflammation.
Catalog No. 50-002-27680 Supplier Medchemexpress LLC Supplier No. HY11825050MG
GSK2245035 is a selective Toll-Like receptor 7 (TLR7) agonist for research use, preferentially stimulating type I interferon production and evaluated for intranasal administration in modulation of allergic responses. It is supplied as a high-purity, off-white to light-yellow solid suitable for laboratory studies.
  • Selective TLR7 agonist with preferential type I interferon stimulation.
  • High purity appropriate for in vitro and in vivo research.
  • Off-white to light-yellow solid form with defined storage stability.
  • Available as powder and as ready-to-use DMSO solutions for flexible dosing.
  • Commonly used in studies of innate immunity, antiviral responses, and allergic inflammation.