Learn More
Selleck Chemical LLC Nemiralisib (GSK2269557) 5mg 1254036-71-9 Nemiralisib
SDP
Supplier:  Selleck Chemical LLC S79375MG
Encompass_Preferred
Nemiralisib, also known as GSK-2269557, is a potent and selective PI3K? inhibitor(pKi = 9.9). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5442