Learn More
Medchemexpress LLC GSK2292767 | 1254036-66-2 | MFCD22572737 | 98.9% | 512.58 | C24H28N6O5S | 10 MG
SDP
Catalog No. 5000285712
Encompass_Preferred
GSK2292767 is a potent, selective PI3Kδ inhibitor supplied for preclinical research use. It is provided as a solid (light yellow to yellow) with reported purity of 98.89% and CAS 1254036-66-2, packaged in small-mass quantities suitable for in vitro and exploratory in vivo studies. Store as powder refrigerated or frozen according to supplier recommendations.
  • Potent and selective PI3Kδ inhibition (pIC50 ~10.1).
  • High reported purity for reliable experimental use.
  • Available in small-mass quantities for screening and dosing studies.
  • Solid form suitable for solution preparation and storage.
  • Documented storage conditions for powder and stock solutions.
Catalog No. 50-002-85712 Supplier Medchemexpress LLC Supplier No. HY1528010MG
GSK2292767 is a potent, selective PI3Kδ inhibitor supplied for preclinical research use. It is provided as a solid (light yellow to yellow) with reported purity of 98.89% and CAS 1254036-66-2, packaged in small-mass quantities suitable for in vitro and exploratory in vivo studies. Store as powder refrigerated or frozen according to supplier recommendations.
  • Potent and selective PI3Kδ inhibition (pIC50 ~10.1).
  • High reported purity for reliable experimental use.
  • Available in small-mass quantities for screening and dosing studies.
  • Solid form suitable for solution preparation and storage.
  • Documented storage conditions for powder and stock solutions.