Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gsk251 | 2125968-05-8 | 99.9% | 584.71 g·mol⁻¹ | C29H37FN6O4S | 5 MG
SDP

Catalog No. 5000262539
Encompass_Preferred

GSK251 is a potent and selective inhibitor of PI3Kδ developed for research use. It is orally bioavailable and supplied as a solid, with published solubility and storage recommendations to support both in vitro and in vivo experiments.

  • High purity: 99.88%.
  • Selective inhibition of PI3Kδ with a novel binding mode.
  • Soluble in DMSO (62.5 mg/mL) and amenable to in vivo formulations.
  • Recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).
  • Available in multiple small pack sizes for screening and dosing studies.

Catalog No. 50-002-62539 Supplier Medchemexpress LLC Supplier No. HY1328805MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GSK251 is a potent and selective inhibitor of PI3Kδ developed for research use. It is orally bioavailable and supplied as a solid, with published solubility and storage recommendations to support both in vitro and in vivo experiments.

  • High purity: 99.88%.
  • Selective inhibition of PI3Kδ with a novel binding mode.
  • Soluble in DMSO (62.5 mg/mL) and amenable to in vivo formulations.
  • Recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).
  • Available in multiple small pack sizes for screening and dosing studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.