Learn More
Medchemexpress LLC GSK251 | 2125968-05-8 | MFCD34602603 | 99.9% | C29H37FN6O4S | 10MG
SDP
Catalog No. 5000259645
Encompass_Preferred
GSK251 is a highly potent, highly selective, orally bioavailable inhibitor of PI3Kδ that binds via a novel binding mode. Provided as a high-purity solid, it is suitable for in vitro and in vivo research investigating PI3Kδ signaling.
  • High purity (99.9%).
  • Selective PI3Kδ inhibition with a novel binding mode.
  • Orally bioavailable profile suitable for in vivo studies.
  • Soluble in DMSO and validated in multiple in vivo formulations.
  • Supplied as an off-white to light yellow solid for ease of handling.
  • Stable when stored under recommended temperature conditions.
Catalog No. 50-002-59645 Supplier Medchemexpress LLC Supplier No. HY13288010MG
GSK251 is a highly potent, highly selective, orally bioavailable inhibitor of PI3Kδ that binds via a novel binding mode. Provided as a high-purity solid, it is suitable for in vitro and in vivo research investigating PI3Kδ signaling.
  • High purity (99.9%).
  • Selective PI3Kδ inhibition with a novel binding mode.
  • Orally bioavailable profile suitable for in vivo studies.
  • Soluble in DMSO and validated in multiple in vivo formulations.
  • Supplied as an off-white to light yellow solid for ease of handling.
  • Stable when stored under recommended temperature conditions.