Learn More
TARGETMOL CHEMICALS INC GSK2593074A 5MG
SDP
Catalog No. 502581527
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GSK2593074A (GSK'074) is a programmed necrosis inhibitor that displays inhibitory effects on RIP1 and RIP3. purity: 100%
Catalog No. 50-258-1527 Supplier TARGETMOL CHEMICALS INC Supplier No. T114845MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GSK2593074A (GSK'074) is a programmed necrosis inhibitor that displays inhibitory effects on RIP1 and RIP3. purity: 100%