Learn More
Medchemexpress LLC HY-18072 10mg , GSK2606414 CAS:1337531-36-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1807210MG
Encompass_Preferred
HY-18072 10mg GSK2606414 CAS: 1337531-36-8 GSK2606414 is a cell-permeable and orally available protein kinase R-like endoplasmic reticulum (ER) kinase (PERK) inhibitor with an IC50 of 0.4 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0213