Learn More
Apexbio Technology LLC GSK2606414, 1337531-36-8, MFCD25976926, 10 mg
SDP
Catalog No. 501015099
Encompass_Preferred
MF: C24H20F3N5O, MW: 451.44, Solubility: 22.57 mg/mL in DMSO, 12.03 mg/mL in EtOH with ultrasonic and warming,insoluble in H2O. GSK2606414 is a selective inhibitor of PERK with IC50 value of 0.4nM.
Catalog No. 50-101-5099 Supplier Apexbio Technology LLC Supplier No. A344810
MF: C24H20F3N5O, MW: 451.44, Solubility: 22.57 mg/mL in DMSO, 12.03 mg/mL in EtOH with ultrasonic and warming,insoluble in H2O. GSK2606414 is a selective inhibitor of PERK with IC50 value of 0.4nM.