Learn More
AdipoGen GSK2606414, 1337531-36-8, MFCD25976926, 1 mg
SDP
Catalog No. 501016107
Encompass_Preferred
MF: C24H20F3N5O, MW: 451.4, ≥95%, Appearance: Solid. GSK2606414 is an orally available, potent, and selective PERK inhibitor with IC(50) of 0.4nM, displaying at least 100-fold selectivity over the other EIF2AKs assayed.
Catalog No. 50-101-6107 Supplier AdipoGen Supplier No. SYN1201M001
MF: C24H20F3N5O, MW: 451.4, ≥95%, Appearance: Solid. GSK2606414 is an orally available, potent, and selective PERK inhibitor with IC(50) of 0.4nM, displaying at least 100-fold selectivity over the other EIF2AKs assayed.