Learn More
Apexbio Technology LLC GSK2606414, 200mg. Cas: 1337531-36-8 MFCD: MFCD25976926. PERK inhibitor,potent and selective
SDP
Supplier:  Apexbio Technology LLC A3448200
Encompass_Preferred
GSK2606414 is a selective inhibitor of PERK with IC50 value of 0.4nM.[1]PERK (PRKR-like endoplasmic reticulum kinase or protein kinase R (PKR)-like endoplasmic reticulum kinase) is also known as EIF2AK3 (Eukaryotic translation initiation factor 2-alpha kinase 3). Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4803