Learn More
Apexbio Technology LLC GSK2606414, 50mg Cas No: 1337531-36-8 MFCD: MFCD25976926
SDP
Catalog No. NC1586131
Encompass_Preferred
GSK2606414 is a selective inhibitor of PERK with IC50 value of 0.4nM.[1]PERK (PRKR-like endoplasmic reticulum kinase or protein kinase R (PKR)-like endoplasmic reticulum kinase) is also known as EIF2AK3 (Eukaryotic translation initiation factor 2-alpha kinase 3). There are other sizes available. Please inqury us for quote.
Catalog No. NC1586131 Supplier Apexbio Technology LLC Supplier No. A344850
GSK2606414 is a selective inhibitor of PERK with IC50 value of 0.4nM.[1]PERK (PRKR-like endoplasmic reticulum kinase or protein kinase R (PKR)-like endoplasmic reticulum kinase) is also known as EIF2AK3 (Eukaryotic translation initiation factor 2-alpha kinase 3). There are other sizes available. Please inqury us for quote.