Learn More
Medchemexpress LLC HY-15245 10mg , GSK2636771 CAS:1372540-25-4 Purity:>98%
SDP
Catalog No. 502259743
Encompass_Preferred
HY-15245 10mg GSK2636771 CAS: 1372540-25-4 GSK2636771 is a potent, selective and orally bioavailable inhibitor of PI3Kβ with a Ki of 0.89 nM and an IC50 of 5.2 nM, showing 900-fold selectivity over p110α and p110γ, and 10-fold selectivity over p110δ isoforms. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9743 Supplier Medchemexpress LLC Supplier No. HY1524510MG
HY-15245 10mg GSK2636771 CAS: 1372540-25-4 GSK2636771 is a potent, selective and orally bioavailable inhibitor of PI3Kβ with a Ki of 0.89 nM and an IC50 of 5.2 nM, showing 900-fold selectivity over p110α and p110γ, and 10-fold selectivity over p110δ isoforms. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.