Learn More
Apexbio Technology LLC GSK2636771, 5mg. Cas: 1372540-25-4 MFCD: MFCD22417098. PIK3 inhibitor
SDP
Supplier:  Apexbio Technology LLC B21865
Encompass_Preferred
GSK2636771 is a p110 isoform-specific inhibitor of PIK3 with SF50 more than 1 M [1].PI3K (phosphatidylinositol 3-kinase) has 4 catalytic subunits: p110, , , and . Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5328