Learn More
Selleck Chemical LLC GSK2636771 5mg 1372540-25-4
SDP
Catalog No. NC0742274
Encompass_Preferred
GSK2636771 is a potent, orally bioavailable and selective inhibitor of PI3K? with >900-fold selectivity over PI3K?/PI3K? and >10-fold over PI3K?. Sensitive to PTEN null cell lines. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0742274 Supplier Selleck Chemical LLC Supplier No. S80025MG
Add to Cart
Add to Cart
GSK2636771 is a potent, orally bioavailable and selective inhibitor of PI3K? with >900-fold selectivity over PI3K?/PI3K? and >10-fold over PI3K?. Sensitive to PTEN null cell lines. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.