Learn More
Medchemexpress LLC HY-111458 10mg Medchemexpress, GSK2643943A CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11145810MG
Encompass_Preferred
Medchemexpress, HY-111458 10mg GSK2643943A CAS: GSK2643943A is a deubiquitylating enzyme (DUB) inhibitor, with an IC50 of 160 nM for USP20/Ub-Rho. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8877