Learn More
Apexbio Technology LLC GSK2656157, 100mg. Cas: 1337532-29-2 MFCD: MFCD27997885. PERK inhibitor
SDP
Supplier:  Apexbio Technology LLC B2175100
Encompass_Preferred
GSK2656157 is a highly selective inhibitor of protein kinase R-like ER kinase (PERK) with IC50 value of 0.9nM [1].GSK2656157 is highly selective for PERK enzyme against a panel of 300 kinases. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5323