Learn More
TARGETMOL CHEMICALS INC GSK2656157 25MG
Supplier: TARGETMOL CHEMICALS INC T265425MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GSK2656157 is a highly specific and ATP-competitive PERK inhibitor (IC50 0.9 nM) in a cell-free assay. The selectivty is 500-fold higher against a panel of 300 kinases. purity: 99%