Learn More
Selleck Chemical LLC GSK2656157 50mg 1337532-29-2
SDP
Catalog No. 501364336
Encompass_Preferred
GSK2656157 is an ATP-competitive and highly selective inhibitor of PERK with IC50 of 0.9 nM in a cell-free assay, 500-fold greater against a panel of 300 kinases. GSK2656157 decreases apoptosis and inhibits excessive autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4336 Supplier Selleck Chemical LLC Supplier No. S703350MG
GSK2656157 is an ATP-competitive and highly selective inhibitor of PERK with IC50 of 0.9 nM in a cell-free assay, 500-fold greater against a panel of 300 kinases. GSK2656157 decreases apoptosis and inhibits excessive autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.