Learn More
Abcam GSK2656157, 25MG
Catalog No. 502671862
Encompass_Preferred
MW 416.5 Da, Purity >=98%. Potent and selective PERK inhibitor (IC₅₀ = 0.9 nM in cell-free assay). ATP-competitive. Exibits 500-fold selectivity for PERK versus a panel of 300 kinases.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-1862 Supplier Abcam Supplier No. AB27362225MG
MW 416.5 Da, Purity >=98%. Potent and selective PERK inhibitor (IC₅₀ = 0.9 nM in cell-free assay). ATP-competitive. Exibits 500-fold selectivity for PERK versus a panel of 300 kinases.
The product is subject to the following: Abcam Restricted Use Statement