Learn More
Medchemexpress LLC HY-18950 10mg , GSK2795039 CAS:1415925-18-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1895010MG
Encompass_Preferred
HY-18950 10mg GSK2795039 CAS: 1415925-18-6 GSK2795039 is a NADPH oxidase 2 (NOX2) inhibitor with a mean pIC50 of 6 in different cell-free assays. GSK2795039 inhibits reactive oxygen species (ROS) production and NADPH consumption.GSK2795039 reduces apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9783