Learn More
Selleck Chemical LLC GSK2801 10mg 1619994-68-1
SDP
Supplier:  Selleck Chemical LLC S723110MG
Encompass_Preferred
GSK2801 is a selective bromodomains BAZ2A/B inhibitor with KD of 257 nM and 136 nM, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1284535