Learn More
Medchemexpress LLC HY-15658 5mg Medchemexpress, GSK2801 CAS:1619994-68-1 Purity:>98%
SDP
Catalog No. 502030662
Encompass_Preferred
Medchemexpress, HY-15658 5mg GSK2801 CAS:1619994-68-1 GSK2801 is a potent, selective, orally active and cell active acetyl-lysine competitive BAZ2A and BAZ2B bromodomains inhibitor with Kd values of 136 nM and 257 nM, respectively. GSK2801 shows >50-fold selectivity for BAZ2A/B over BRD4. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0662 Supplier Medchemexpress LLC Supplier No. HY156585MG
Add to Cart
Add to Cart
Medchemexpress, HY-15658 5mg GSK2801 CAS:1619994-68-1 GSK2801 is a potent, selective, orally active and cell active acetyl-lysine competitive BAZ2A and BAZ2B bromodomains inhibitor with Kd values of 136 nM and 257 nM, respectively. GSK2801 shows >50-fold selectivity for BAZ2A/B over BRD4. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.