Learn More
Medchemexpress LLC GSK2807 trifluoroacetate | 2245255-66-5 | 99.8% | 566.53 | C21H33F3N8O7 | 100 MG
SDP
Catalog No. 5000241890
Encompass_Preferred
GSK2807 trifluoroacetate is a SAM-competitive small-molecule inhibitor of the histone methyltransferase SMYD3, reported with Ki = 14 nM and IC50 = 130 nM. It is provided as the trifluoroacetate salt for research use in biochemical and cell-based studies of methylation-dependent signaling.
  • High potency against SMYD3 (Ki = 14 nM; IC50 = 130 nM).
  • SAM-competitive mechanism of action.
  • Supplied as trifluoroacetate (TFA) salt.
  • High chemical purity (>99.8%).
  • Molecular weight 566.53 g/mol.
  • Suitable for biochemical and cell-based assay development.
Catalog No. 50-002-41890 Supplier Medchemexpress LLC Supplier No. HY104009A100MG
GSK2807 trifluoroacetate is a SAM-competitive small-molecule inhibitor of the histone methyltransferase SMYD3, reported with Ki = 14 nM and IC50 = 130 nM. It is provided as the trifluoroacetate salt for research use in biochemical and cell-based studies of methylation-dependent signaling.
  • High potency against SMYD3 (Ki = 14 nM; IC50 = 130 nM).
  • SAM-competitive mechanism of action.
  • Supplied as trifluoroacetate (TFA) salt.
  • High chemical purity (>99.8%).
  • Molecular weight 566.53 g/mol.
  • Suitable for biochemical and cell-based assay development.