Learn More
Medchemexpress LLC Gsk2807 trifluoroacetate | 2245255-66-5 | 99.8% | 566.53 g·mol⁻¹ | C21H33F3N8O7 | 10 MM 1 ML
SDP
Catalog No. 5000243895
Encompass_Preferred
GSK2807 trifluoroacetate is the trifluoroacetate salt form of a potent, selective SAM-competitive inhibitor of the histone methyltransferase SMYD3. Supplied as a ready-to-use 10 mM solution in DMSO, it is intended for research applications in epigenetic modulation and target validation. Purity is 99.8%, and documentation including COA and SDS is available.
  • Potent, selective SAM-competitive SMYD3 inhibition with nanomolar activity.
  • Trifluoroacetate salt form enhances solubility and handling for assays.
  • Supplied as a 10 mM solution in DMSO for immediate use.
  • High purity with supporting COA and SDS documentation.
  • Suitable for biochemical assays, cellular studies, and target validation.
Catalog No. 50-002-43895 Supplier Medchemexpress LLC Supplier No. HY104009A10MM 1ML
GSK2807 trifluoroacetate is the trifluoroacetate salt form of a potent, selective SAM-competitive inhibitor of the histone methyltransferase SMYD3. Supplied as a ready-to-use 10 mM solution in DMSO, it is intended for research applications in epigenetic modulation and target validation. Purity is 99.8%, and documentation including COA and SDS is available.
  • Potent, selective SAM-competitive SMYD3 inhibition with nanomolar activity.
  • Trifluoroacetate salt form enhances solubility and handling for assays.
  • Supplied as a 10 mM solution in DMSO for immediate use.
  • High purity with supporting COA and SDS documentation.
  • Suitable for biochemical assays, cellular studies, and target validation.