Learn More
Medchemexpress LLC GSK2807 trifluoroacetate | 2245255-66-5 | 99.8% | 566.53 g·mol⁻¹ | C21H33F3N8O7 | 25 MG
SDP
Catalog No. 5000241878
Encompass_Preferred
GSK2807 Trifluoroacetate is the trifluoroacetate salt of a potent, selective, SAM-competitive inhibitor of the histone methyltransferase SMYD3. Supplied as a high-purity solid for research use, it demonstrates biochemical potency (Ki 14 nM; IC50 130 nM) and includes manufacturer guidance for solubility and storage for in vitro and in vivo applications.
  • Potent, selective SAM-competitive inhibition of SMYD3 (Ki 14 nM, IC50 130 nM).
  • High purity solid suitable for biochemical and cellular studies.
  • Formulated as a trifluoroacetate salt to improve handling and stability.
  • Well-characterized solubility in DMSO and water for assay preparation.
  • Storage guidance provided for refrigerated and long-term frozen conditions.
Catalog No. 50-002-41878 Supplier Medchemexpress LLC Supplier No. HY104009A25MG
GSK2807 Trifluoroacetate is the trifluoroacetate salt of a potent, selective, SAM-competitive inhibitor of the histone methyltransferase SMYD3. Supplied as a high-purity solid for research use, it demonstrates biochemical potency (Ki 14 nM; IC50 130 nM) and includes manufacturer guidance for solubility and storage for in vitro and in vivo applications.
  • Potent, selective SAM-competitive inhibition of SMYD3 (Ki 14 nM, IC50 130 nM).
  • High purity solid suitable for biochemical and cellular studies.
  • Formulated as a trifluoroacetate salt to improve handling and stability.
  • Well-characterized solubility in DMSO and water for assay preparation.
  • Storage guidance provided for refrigerated and long-term frozen conditions.