Learn More
Medchemexpress LLC GSK2807 trifluoroacetate | 2245255-66-5 | 99.8% | 566.53 g/mol | C21H33F3N8O7 | 50 MG
SDP
Catalog No. 5000243896
Encompass_Preferred
GSK2807 trifluoroacetate is a selective, SAM-competitive inhibitor of the protein methyltransferase SMYD3 with potent biochemical activity (Ki = 14 nM; IC50 = 130 nM). Supplied as the trifluoroacetate salt and formulated for research applications, it is intended for biochemical and cellular studies of SMYD3-mediated methylation and epigenetic regulation.
  • Provided as a trifluoroacetate salt for improved handling
  • Potent SMYD3 inhibition (Ki 14 nM; IC50 130 nM)
  • High purity suitable for research use
  • Compact 50 mg package size for assay development
  • Well characterized for biochemical and cellular studies
Catalog No. 50-002-43896 Supplier Medchemexpress LLC Supplier No. HY104009A50MG
GSK2807 trifluoroacetate is a selective, SAM-competitive inhibitor of the protein methyltransferase SMYD3 with potent biochemical activity (Ki = 14 nM; IC50 = 130 nM). Supplied as the trifluoroacetate salt and formulated for research applications, it is intended for biochemical and cellular studies of SMYD3-mediated methylation and epigenetic regulation.
  • Provided as a trifluoroacetate salt for improved handling
  • Potent SMYD3 inhibition (Ki 14 nM; IC50 130 nM)
  • High purity suitable for research use
  • Compact 50 mg package size for assay development
  • Well characterized for biochemical and cellular studies