Learn More
Medchemexpress LLC GSK2807 trifluoroacetate | 2245255-66-5 | MFCD31715435 | 99.8% | 566.53 g·mol⁻¹ | C21H33F3N8O7 | 10 MG
SDP
Catalog No. 5000242650
Encompass_Preferred
GSK2807 trifluoroacetate is the trifluoroacetate salt of a potent, selective, SAM-competitive inhibitor of the histone methyltransferase SMYD3 (Ki = 14 nM; IC50 = 130 nM). It is supplied as a high-purity research compound for biochemical and cellular studies.
  • Potent SAM-competitive inhibition of SMYD3 (Ki 14 nM; IC50 130 nM).
  • High purity (~99.8%) suitable for biochemical and cellular assays.
  • Provided as the trifluoroacetate salt to aid solubility and handling.
  • Available in small milligram quantities and as a DMSO solution for screening.
  • Recommended storage: solid at 4°C; solutions at -80°C (up to 6 months) or -20°C (up to 1 month).
Catalog No. 50-002-42650 Supplier Medchemexpress LLC Supplier No. HY104009A10MG
GSK2807 trifluoroacetate is the trifluoroacetate salt of a potent, selective, SAM-competitive inhibitor of the histone methyltransferase SMYD3 (Ki = 14 nM; IC50 = 130 nM). It is supplied as a high-purity research compound for biochemical and cellular studies.
  • Potent SAM-competitive inhibition of SMYD3 (Ki 14 nM; IC50 130 nM).
  • High purity (~99.8%) suitable for biochemical and cellular assays.
  • Provided as the trifluoroacetate salt to aid solubility and handling.
  • Available in small milligram quantities and as a DMSO solution for screening.
  • Recommended storage: solid at 4°C; solutions at -80°C (up to 6 months) or -20°C (up to 1 month).