Learn More
TARGETMOL CHEMICALS INC GSK2807 Trifluoroacetate 5MG
SDP
Catalog No. 502880561
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. GSK2807 Trifluoroacetate is a selective and SAM-competitive inhibitor of SMYD3 (Ki 14 nM; IC50 130 nM). Purity 99.95%
Catalog No. 50-288-0561 Supplier TARGETMOL CHEMICALS INC Supplier No. T114865MG
Also available in 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. GSK2807 Trifluoroacetate is a selective and SAM-competitive inhibitor of SMYD3 (Ki 14 nM; IC50 130 nM). Purity 99.95%