Learn More
Medchemexpress LLC HY-104009A 5mg Medchemexpress, GSK2807 Trifluoroacetate CAS:2245255-66-5 Purity:>98%
SDP
Catalog No. 501968185
Encompass_Preferred
Medchemexpress, HY-104009A 5mg GSK2807 Trifluoroacetate CAS:2245255-66-5 GSK2807 Trifluoroacetate is a potent, selective and SAM-competitive inhibitor of SMYD3, with a Ki of 14 nM and an IC50 of 130 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8185 Supplier Medchemexpress LLC Supplier No. HY104009A5MG
Medchemexpress, HY-104009A 5mg GSK2807 Trifluoroacetate CAS:2245255-66-5 GSK2807 Trifluoroacetate is a potent, selective and SAM-competitive inhibitor of SMYD3, with a Ki of 14 nM and an IC50 of 130 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.