Learn More
Selleck Chemical LLC GSK2830371 10mg 1404456-53-6
SDP
Catalog No. 501364961
Encompass_Preferred
GSK2830371 is an orally active, allosteric Wip1 phosphatase inhibitor with IC50 of 6 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4961 Supplier Selleck Chemical LLC Supplier No. S757310MG
GSK2830371 is an orally active, allosteric Wip1 phosphatase inhibitor with IC50 of 6 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.