Learn More
AdipoGen GSK2837808A (1 mg)
SDP
Supplier:  AdipoGen AGCR13685M001
Encompass_Preferred
GSK2837808A (1 mg), Adipogen Life Sciences. Chemical. CAS: 1445879-21-9. Formula: C31H25F2N5O7S. MW: 649.6. . Potent and selective cell-permeable inhibitor of lactate dehydrogenase A (LDHA) (IC50 values are 2.6 and 43 nM for LDHA and LDHB, respectively). Decreases glycolysis. Useful agent for immunometabolism research. Exhibits antitumoral activity. Inhibits lactate production in selected cancer cell lines. Reduces glucose uptake and enhances mitochondrial oxygen consumption in Snu398 hepatocellular carcinoma cells. Inhibits proliferation and induces apoptosis in Snu398 cells. Inhibits transcription of histone 2B (H2B) gene in HCT116 and NCM460 cells. Lactate dehydrogenase A (LDHA) is an important enzyme involved in fermentative glycolysis, important in tumor initiation and metabolism. Cancer cells rely on increased glycolysis resulting in increased lactate production in addition to aerobic respiration in the mitochondria, even under normal oxygen concentrations (Warburg effect).
Catalog No. 50-168-7448