Learn More
Medchemexpress LLC HY-100681 10mg Medchemexpress, GSK2837808A CAS:1445879-21-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10068110MG
Encompass_Preferred
Medchemexpress, HY-100681 10mg GSK2837808A CAS:1445879-21-9 GSK2837808A is a potent and selective lactate dehydrogenase A (LDHA) inhibitor with IC50s of 1.9 and 14 nM for LDHA and LDHB, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7767