Learn More
AdipoGen GSK2837808A
SDP
Supplier:  AdipoGen AGCR13685M005
Encompass_Preferred
Chemical. CAS 1445879-21-9. Formula C31H25F2N5O7S. MW 649.6. . Potent and selective cell-permeable inhibitor of lactate dehydrogenase A LDHA IC50 values are 2.6 and 43 nM for LDHA and LDHB, respectively. Decreases glycolysis. Useful agent for immunometabolism research. Exhibits antitumoral activity. Inhibits lactate production in selected cancer cell lines. Reduces glucose uptake and enhances mitochondrial oxygen consumption in Snu398 hepatocellular carcinoma cells. Inhibits proliferation and induces apoptosis in Snu398 cells. Inhibits transcription of histone 2B H2B gene in HCT116 and NCM460 cells. Lactate dehydrogenase A LDHA is an important enzyme involved in fermentative glycolysis, important in tumor initiation and metabolism. Cancer cells rely on increased glycolysis resulting in increased lactate production in addition to aerobic respiration in the mitochondria, even under normal oxygen concentrations a process known as the Warburg effect.
Catalog No. 50-256-2227