Learn More
Cayman Chemical GSK2837808A, Purity: ≥98%, CAS: 1445879-21-9, 5 mg
SDP
Catalog No. 502982274
Encompass_Preferred
A potent inhibitor of LDHA and LDHB (IC50s = 2.6 and 0.43 nM, for human recombinant LDHA and LDHB, respectively); inhibits lactate production in Snu398 hepatocellular carcinoma cells in which LDHB expression is undetectable (EC50 = 400 nM); reduces glucose consumption in Snu398 but not HepG2 cells; 30 cancer cell lines show differential sensitivities to GSK2837808A (EC50s from 400 nM to 30 µM); potency does not correlate with LDHA, LDHB, or total LDH expression levels
Catalog No. 50-298-2274 Supplier Cayman Chemical Supplier No. 206265 MG
A potent inhibitor of LDHA and LDHB (IC50s = 2.6 and 0.43 nM, for human recombinant LDHA and LDHB, respectively); inhibits lactate production in Snu398 hepatocellular carcinoma cells in which LDHB expression is undetectable (EC50 = 400 nM); reduces glucose consumption in Snu398 but not HepG2 cells; 30 cancer cell lines show differential sensitivities to GSK2837808A (EC50s from 400 nM to 30 µM); potency does not correlate with LDHA, LDHB, or total LDH expression levels