Learn More
Medchemexpress LLC HY-U00459 5mg Medchemexpress, GSK2850163 CAS:2121989-91-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYU004595MG
Encompass_Preferred
Medchemexpress, HY-U00459 5mg GSK2850163 CAS:2121989-91-9 GSK2850163 is a novel inhibitor of inositol-requiring enzyme-1 alpha ( IRE1α ) which can inhibit IRE1α kinase activity and RNase activity with IC 50 s of 20 and 200 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4459