Learn More
TARGETMOL CHEMICALS INC GSK2850163 S ENANTIOMER 5MG
SDP
Catalog No. 502578950
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GSK2850163 (S enantiomer) is the inactive enantiomer of GSK2850163. GSK2850163 is an novel inhibitor of inositol-requiring enzyme-1 alpha (IRE1a). purity: 98%
Catalog No. 50-257-8950 Supplier TARGETMOL CHEMICALS INC Supplier No. T98485MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GSK2850163 (S enantiomer) is the inactive enantiomer of GSK2850163. GSK2850163 is an novel inhibitor of inositol-requiring enzyme-1 alpha (IRE1a). purity: 98%